Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 450, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 450, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 450, eBioscience™ - FAQs (48-4015-80, 48-4015-82)