eBioscience™ Streptavidin eFluor™ 450 Conjugate - FAQs
eBioscience™ Streptavidin eFluor™ 450 Conjugate - FAQs
View additional product information for eBioscience™ Streptavidin eFluor™ 450 Conjugate - FAQs (48-4317-82)