Rat IgM Isotype Control (eBRM), eFluor™ 450, eBioscience™ - FAQs
Rat IgM Isotype Control (eBRM), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Rat IgM Isotype Control (eBRM), eFluor™ 450, eBioscience™ - FAQs (48-4341-80)