Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 450, eBioscience™ - FAQs
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 450, eBioscience™ - FAQs (48-4724-82, 48-4724-80)