Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 450, eBioscience™ - FAQs (48-4732-82, 48-4732-80)