Mouse IgM Isotype Control (11E10), eFluor™ 450, eBioscience™ - FAQs
Mouse IgM Isotype Control (11E10), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgM Isotype Control (11E10), eFluor™ 450, eBioscience™ - FAQs (48-4752-80)