FOXP3 Monoclonal Antibody (PCH101), eFluor™ 450, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (PCH101), eFluor™ 450, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (PCH101), eFluor™ 450, eBioscience™ - FAQs (48-4776-42, 48-4776-41)