FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 450, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 450, eBioscience™ - FAQs (48-4777-42, 48-4777-41)