F4/80 Monoclonal Antibody (BM8), eFluor™ 450, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), eFluor™ 450, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), eFluor™ 450, eBioscience™ - FAQs (48-4801-80, 48-4801-82)