EOMES Monoclonal Antibody (Dan11mag), eFluor™ 450, eBioscience™ - FAQs
EOMES Monoclonal Antibody (Dan11mag), eFluor™ 450, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (Dan11mag), eFluor™ 450, eBioscience™ - FAQs (48-4875-80, 48-4875-82)