Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 450, eBioscience™ - FAQs
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 450, eBioscience™ - FAQs (48-4888-82, 48-4888-80)