Syrian Hamster IgG Isotype Control, eFluor™ 450, eBioscience™ - FAQs
Syrian Hamster IgG Isotype Control, eFluor™ 450, eBioscience™ - FAQs
View additional product information for Syrian Hamster IgG Isotype Control, eFluor™ 450, eBioscience™ - FAQs (48-4914-80)