phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 450, eBioscience™ - FAQs
phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 450, eBioscience™ - FAQs
View additional product information for phospho-Tyrosine Monoclonal Antibody (pY20), eFluor™ 450, eBioscience™ - FAQs (48-5001-42, 48-5001-41)