BrdU Monoclonal Antibody (BU20A), eFluor™ 450, eBioscience™ - FAQs
BrdU Monoclonal Antibody (BU20A), eFluor™ 450, eBioscience™ - FAQs
View additional product information for BrdU Monoclonal Antibody (BU20A), eFluor™ 450, eBioscience™ - FAQs (48-5071-42, 48-5071-41)