CD45 Monoclonal Antibody (YKIX716.13), eFluor™ 450, eBioscience™ - FAQs
CD45 Monoclonal Antibody (YKIX716.13), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (YKIX716.13), eFluor™ 450, eBioscience™ - FAQs (48-5450-42, 48-5450-41)