Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 450, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 450, eBioscience™ - FAQs (48-5699-42, 48-5699-41)