FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 450, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 450, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 450, eBioscience™ - FAQs (48-5773-80, 48-5773-82)