Mouse IgM Monoclonal Antibody (II/41), eFluor™ 450, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), eFluor™ 450, eBioscience™ - FAQs (48-5790-82, 48-5790-80)