CD326 (EpCAM) Monoclonal Antibody (G8.8), eFluor™ 450, eBioscience™ - FAQs
CD326 (EpCAM) Monoclonal Antibody (G8.8), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD326 (EpCAM) Monoclonal Antibody (G8.8), eFluor™ 450, eBioscience™ - FAQs (48-5791-82, 48-5791-80)