TCR V alpha 24 J alpha 18 Monoclonal Antibody (6B11), eFluor™ 450, eBioscience™ - FAQs
TCR V alpha 24 J alpha 18 Monoclonal Antibody (6B11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TCR V alpha 24 J alpha 18 Monoclonal Antibody (6B11), eFluor™ 450, eBioscience™ - FAQs (48-5806-42, 48-5806-41)