TCR V alpha 2 Monoclonal Antibody (B20.1), eFluor™ 450, eBioscience™ - FAQs
TCR V alpha 2 Monoclonal Antibody (B20.1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TCR V alpha 2 Monoclonal Antibody (B20.1), eFluor™ 450, eBioscience™ - FAQs (48-5812-82, 48-5812-80)