TCR V beta 9 Monoclonal Antibody (MR10-2), eFluor™ 450, eBioscience™ - FAQs
TCR V beta 9 Monoclonal Antibody (MR10-2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TCR V beta 9 Monoclonal Antibody (MR10-2), eFluor™ 450, eBioscience™ - FAQs (48-5823-80)