Granzyme A Monoclonal Antibody (GzA-3G8.5), eFluor™ 450, eBioscience™ - FAQs
Granzyme A Monoclonal Antibody (GzA-3G8.5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Granzyme A Monoclonal Antibody (GzA-3G8.5), eFluor™ 450, eBioscience™ - FAQs (48-5831-82, 48-5831-80)