CD366 (TIM3) Monoclonal Antibody (8B.2C12), eFluor™ 450, eBioscience™ - FAQs
CD366 (TIM3) Monoclonal Antibody (8B.2C12), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD366 (TIM3) Monoclonal Antibody (8B.2C12), eFluor™ 450, eBioscience™ - FAQs (48-5871-80, 48-5871-82)