Mouse IgM Monoclonal Antibody (eB121-15F9), eFluor™ 450, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (eB121-15F9), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (eB121-15F9), eFluor™ 450, eBioscience™ - FAQs (48-5890-80, 48-5890-82)