CD93 (AA4.1) Monoclonal Antibody (AA4.1), eFluor™ 450, eBioscience™ - FAQs
CD93 (AA4.1) Monoclonal Antibody (AA4.1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD93 (AA4.1) Monoclonal Antibody (AA4.1), eFluor™ 450, eBioscience™ - FAQs (48-5892-80)