KLRG1 Monoclonal Antibody (2F1), eFluor™ 450, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (2F1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (2F1), eFluor™ 450, eBioscience™ - FAQs (48-5893-80)