TER-119 Monoclonal Antibody (TER-119), eFluor™ 450, eBioscience™ - FAQs
TER-119 Monoclonal Antibody (TER-119), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TER-119 Monoclonal Antibody (TER-119), eFluor™ 450, eBioscience™ - FAQs (48-5921-82, 48-5921-80)