Ly-6C Monoclonal Antibody (HK1.4), eFluor™ 450, eBioscience™ - FAQs
Ly-6C Monoclonal Antibody (HK1.4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Ly-6C Monoclonal Antibody (HK1.4), eFluor™ 450, eBioscience™ - FAQs (48-5932-80, 48-5932-82)