NK1.1 Monoclonal Antibody (PK136), eFluor™ 450, eBioscience™ - FAQs
NK1.1 Monoclonal Antibody (PK136), eFluor™ 450, eBioscience™ - FAQs
View additional product information for NK1.1 Monoclonal Antibody (PK136), eFluor™ 450, eBioscience™ - FAQs (48-5941-80, 48-5941-82)