TCR beta Monoclonal Antibody (H57-597), eFluor™ 450, eBioscience™ - FAQs
TCR beta Monoclonal Antibody (H57-597), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TCR beta Monoclonal Antibody (H57-597), eFluor™ 450, eBioscience™ - FAQs (48-5961-82, 48-5961-80)