Ly-6D Monoclonal Antibody (49-H4), eFluor™ 450, eBioscience™ - FAQs
Ly-6D Monoclonal Antibody (49-H4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Ly-6D Monoclonal Antibody (49-H4), eFluor™ 450, eBioscience™ - FAQs (48-5974-80)