Bcl-2 Monoclonal Antibody (10C4), eFluor™ 450, eBioscience™ - FAQs
Bcl-2 Monoclonal Antibody (10C4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Bcl-2 Monoclonal Antibody (10C4), eFluor™ 450, eBioscience™ - FAQs (48-6992-42, 48-6992-41)