IL-2 Monoclonal Antibody (JES6-5H4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-2 Monoclonal Antibody (JES6-5H4), eFluor™ 450, eBioscience™ - FAQs (48-7021-82, 48-7021-80)