IL-6 Monoclonal Antibody (MQ2-13A5), eFluor™ 450, eBioscience™ - FAQs
IL-6 Monoclonal Antibody (MQ2-13A5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-6 Monoclonal Antibody (MQ2-13A5), eFluor™ 450, eBioscience™ - FAQs (48-7069-42, 48-7069-41)