IL-9 Monoclonal Antibody (MH9D1), eFluor™ 450, eBioscience™ - FAQs
IL-9 Monoclonal Antibody (MH9D1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-9 Monoclonal Antibody (MH9D1), eFluor™ 450, eBioscience™ - FAQs (48-7098-41, 48-7098-42)