IL-10 Monoclonal Antibody (JES3-9D7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-10 Monoclonal Antibody (JES3-9D7), eFluor™ 450, eBioscience™ - FAQs (48-7108-41, 48-7108-42)