IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 450, eBioscience™ - FAQs
IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 450, eBioscience™ - FAQs (48-7129-42, 48-7129-41)