IL-17F Monoclonal Antibody (SHLR17), eFluor™ 450, eBioscience™ - FAQs
IL-17F Monoclonal Antibody (SHLR17), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-17F Monoclonal Antibody (SHLR17), eFluor™ 450, eBioscience™ - FAQs (48-7169-42, 48-7169-41)