IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 450, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 450, eBioscience™ - FAQs (48-7177-82, 48-7177-80)