IL-17A Monoclonal Antibody (eBio64DEC17), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio64DEC17), eFluor™ 450, eBioscience™ - FAQs (48-7179-41, 48-7179-42)