IFN gamma Monoclonal Antibody (XMG1.2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (XMG1.2), eFluor™ 450, eBioscience™ - FAQs (48-7311-82, 48-7311-80)