IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 450, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 450, eBioscience™ - FAQs (48-7319-42, 48-7319-41)