TNF alpha Monoclonal Antibody (MP6-XT22), eFluor™ 450, eBioscience™ - FAQs
TNF alpha Monoclonal Antibody (MP6-XT22), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TNF alpha Monoclonal Antibody (MP6-XT22), eFluor™ 450, eBioscience™ - FAQs (48-7321-80, 48-7321-82)