TNF alpha Monoclonal Antibody (MAb11), eFluor™ 450, eBioscience™ - FAQs
TNF alpha Monoclonal Antibody (MAb11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TNF alpha Monoclonal Antibody (MAb11), eFluor™ 450, eBioscience™ - FAQs (48-7349-42, 48-7349-41)