Granzyme B Monoclonal Antibody (N4TL33), eFluor™ 450, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (N4TL33), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (N4TL33), eFluor™ 450, eBioscience™ - FAQs (48-8896-41, 48-8896-42)