Granzyme B Monoclonal Antibody (NGZB), eFluor™ 450, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (NGZB), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (NGZB), eFluor™ 450, eBioscience™ - FAQs (48-8898-80, 48-8898-82)