Phospho-STAT1 (Tyr701) Monoclonal Antibody (KIKSI0803), eFluor™ 450, eBioscience™ - FAQs
Phospho-STAT1 (Tyr701) Monoclonal Antibody (KIKSI0803), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Phospho-STAT1 (Tyr701) Monoclonal Antibody (KIKSI0803), eFluor™ 450, eBioscience™ - FAQs (48-9008-41, 48-9008-42)