IL-17AF Monoclonal Antibody (20LJS09), eFluor™ 450, eBioscience™ - FAQs
IL-17AF Monoclonal Antibody (20LJS09), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-17AF Monoclonal Antibody (20LJS09), eFluor™ 450, eBioscience™ - FAQs (48-9179-42)