ID2 Monoclonal Antibody (ILCID2), eFluor™ 450, eBioscience™ - FAQs
ID2 Monoclonal Antibody (ILCID2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for ID2 Monoclonal Antibody (ILCID2), eFluor™ 450, eBioscience™ - FAQs (48-9475-82, 48-9475-80)